ENA321763831 Enamine

2-(2-chlorophenoxy)pyridin-4-amine

  • MDL number MFCD16084399
  • Cas Number : 1250565-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75736
Availability details for
Quantity
  • Availability
  • Add to Cart