ENA321763835 Enamine

3-(2-methoxyphenyl)-2,2-dimethylpropanoic acid

  • MDL number MFCD16307198
  • Cas Number : 566157-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44...46
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75741

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75741
Availability details for
Quantity
  • Availability
  • Add to Cart