ENA321763837 Enamine

1-(2-cyclopropyl-1,3-thiazol-4-yl)ethan-1-amine

  • MDL number MFCD16481376
  • Cas Number : 1225632-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75744
Availability details for
Quantity
  • Availability
  • Add to Cart