ENA321763839 Enamine

ethyl 4-oxo-4-(pyridin-3-yl)butanoate

  • MDL number MFCD01320356
  • Cas Number : 59086-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75746

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75746
Availability details for
Quantity
  • Availability
  • Add to Cart