ENA321763840 Enamine

4-(3-bromopropoxy)benzonitrile

  • MDL number MFCD11102045
  • Cas Number : 37136-86-0
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74697
Availability details for
Quantity
  • Availability
  • Add to Cart