ENA321763842 Enamine

3-(3-phenylpropoxy)benzaldehyde

  • MDL number MFCD09712117
  • Cas Number : 938314-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74698
Availability details for
Quantity
  • Availability
  • Add to Cart