ENA321763845 Enamine

2-amino-N-(2-methoxyethyl)acetamide hydrochloride

  • MDL number MFCD08060118
  • Cas Number : 1220037-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74702

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74702
Availability details for
Quantity
  • Availability
  • Add to Cart