ENA321763850 Enamine

2-(2-{[(tert-butoxy)carbonyl]amino}ethoxy)acetic acid

  • MDL number MFCD09909935
  • Cas Number : 142929-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74705

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-74705
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart