ENA321763851 Enamine

N-(3-hydroxyphenyl)-2-methylpropanamide

  • MDL number MFCD01695805
  • Cas Number : 55791-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75784

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75784
Availability details for
Quantity
  • Availability
  • Add to Cart