ENA321763855 Enamine

oxolane-3-sulfonyl chloride

  • MDL number MFCD16090164
  • Cas Number : 1207346-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75790

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-75790
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart