ENA321763856 Enamine

[2-(morpholin-4-yl)-1,3-thiazol-5-yl]methanol

  • MDL number MFCD09879935
  • Cas Number : 921938-89-8
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74349
Availability details for
Quantity
  • Availability
  • Add to Cart