ENA321763861 Enamine

hexane-2-sulfonamide

  • MDL number MFCD16672076
  • Cas Number : 1248031-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75796

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75796
Availability details for
Quantity
  • Availability
  • Add to Cart