ENA321763872 Enamine

1-[2-(4-chlorophenyl)-2-oxoethyl]piperidine-4-carboxamide

  • MDL number MFCD09166230
  • Cas Number : 946817-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74352
Availability details for
Quantity
  • Availability
  • Add to Cart