ENA321763876 Enamine

2-(diethylsulfamoyl)acetic acid

  • MDL number MFCD11107714
  • Cas Number : 956723-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75819
Availability details for
Quantity
  • Availability
  • Add to Cart