ENA321763877 Enamine

(1R)-1-(furan-2-yl)ethan-1-ol

  • MDL number MFCD00077773
  • Cas Number : 27948-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75820

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-75820
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart