ENA321763882 Enamine

tert-butyl N-(2-cyanoethyl)carbamate

  • MDL number MFCD02180880
  • Cas Number : 53588-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-45
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74733
Availability details for
Quantity
  • Availability
  • Add to Cart