ENA321763884 Enamine

N-(3-hydroxyphenyl)propane-2-sulfonamide

  • MDL number MFCD12799091
  • Cas Number : 1179957-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74734
Availability details for
Quantity
  • Availability
  • Add to Cart