ENA321763887 Enamine

2-chloro-6-(methylamino)pyridine-4-carboxylic acid

  • MDL number MFCD11214867
  • Cas Number : 56835-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74737
Availability details for
Quantity
  • Availability
  • Add to Cart