ENA321763895 Enamine

5-nitroquinoline-8-carboxylic acid

  • MDL number MFCD01435910
  • Cas Number : 83848-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75852

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-75852
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart