ENA321763896 Enamine

2-tert-butyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD11847680
  • Cas Number : 42351-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-148
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75853
Availability details for
Quantity
  • Availability
  • Add to Cart