ENA321763898 Enamine

3-(furan-2-yl)-2-(phenylformamido)prop-2-enoic acid

  • MDL number MFCD01817624
  • Cas Number : 70984-54-2
  • Molecular Weight : 257.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75856
Availability details for
Quantity
  • Availability
  • Add to Cart