ENA321763908 Enamine

2-(2-aminophenyl)-5-bromo-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD12569834
  • Cas Number : 1098352-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75869
Availability details for
Quantity
  • Availability
  • Add to Cart