ENA321763912 Enamine

(4-ethoxy-3-methylphenyl)methanamine

  • MDL number MFCD12766962
  • Cas Number : 1184137-77-6
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74354
Availability details for
Quantity
  • Availability
  • Add to Cart