ENA321763914 Enamine

2-methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-7-ol

  • MDL number MFCD16679018
  • Cas Number : 134767-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80...82
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75880

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75880
Availability details for
Quantity
  • Availability
  • Add to Cart