ENA321763915 Enamine

2-{hydroxy[(3-hydroxypropyl)amino]methylidene}-2,3-dihydropyridin-3-one

  • Cas Number : 1281691-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74749
Availability details for
Quantity
  • Availability
  • Add to Cart