ENA321763916 Enamine

4-[(3-methoxypropyl)sulfanyl]aniline

  • MDL number MFCD12768288
  • Cas Number : 1182760-05-9
  • Molecular Weight : 197.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75882
Availability details for
Quantity
  • Availability
  • Add to Cart