ENA321763917 Enamine

3-(methoxymethyl)phenol

  • MDL number MFCD14680990
  • Cas Number : 57234-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75883
Availability details for
Quantity
  • Availability
  • Add to Cart