ENA321763921 Enamine

2-[(3-fluorophenyl)amino]acetonitrile

  • MDL number MFCD11152926
  • Cas Number : 64204-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75889
Availability details for
Quantity
  • Availability
  • Add to Cart