ENA321763922 Enamine

N-methyl-4-(methylsulfanyl)aniline

  • MDL number MFCD09743426
  • Cas Number : 58259-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75890
Availability details for
Quantity
  • Availability
  • Add to Cart