ENA321763924 Enamine

3-(aminomethyl)-4-fluorobenzene-1-sulfonamide

  • MDL number MFCD17167261
  • Cas Number : 1258651-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75892
Availability details for
Quantity
  • Availability
  • Add to Cart