ENA321763925 Enamine

2-(2,3-dihydro-1H-inden-1-ylamino)ethan-1-ol

  • MDL number MFCD10690646
  • Cas Number : 99858-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75893

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75893
Availability details for
Quantity
  • Availability
  • Add to Cart