ENA321763928 Enamine

1-(2-chlorophenyl)-2-methylpropan-1-one

  • MDL number MFCD12824575
  • Cas Number : 5408-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75896
Availability details for
Quantity
  • Availability
  • Add to Cart