ENA321763934 Enamine

1-cyclohexyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09965722
  • Cas Number : 1177299-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74753

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-74753
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart