ENA321763935 Enamine

2-[(4-oxopentyl)oxy]benzamide

  • MDL number MFCD12058665
  • Cas Number : 1157543-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74754
Availability details for
Quantity
  • Availability
  • Add to Cart