ENA321763940 Enamine

2-(1-aminoethyl)-4-fluoro-N,N-dimethylaniline

  • MDL number MFCD11137153
  • Cas Number : 1019602-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74358
Availability details for
Quantity
  • Availability
  • Add to Cart