ENA321763944 Enamine

3-benzylazetidine hydrochloride

  • MDL number MFCD16619075
  • Cas Number : 7606-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74761
Availability details for
Quantity
  • Availability
  • Add to Cart