ENA321763947 Enamine

1-acetyl-2,3-dihydro-1H-indole-6-sulfonyl chloride

  • MDL number MFCD18838573
  • Cas Number : 313690-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75928
Availability details for
Quantity
  • Availability
  • Add to Cart