ENA321763949 Enamine

N,N-dimethyl-2,3-dihydro-1H-indole-6-sulfonamide

  • MDL number MFCD18914704
  • Cas Number : 155110-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75930
Availability details for
Quantity
  • Availability
  • Add to Cart