ENA321763951 Enamine

1-(2-oxopropyl)-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione

  • MDL number MFCD18838730
  • Cas Number : 57384-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75933
Availability details for
Quantity
  • Availability
  • Add to Cart