ENA321763953 Enamine

1-(4,5-dimethyl-1H-imidazol-1-yl)propan-2-one

  • MDL number MFCD16715808
  • Cas Number : 1250237-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80...82
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75937

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75937
Availability details for
Quantity
  • Availability
  • Add to Cart