ENA321763957 Enamine

2-(2-hydroxyethoxy)benzamide

  • MDL number MFCD00025473
  • Cas Number : 63906-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75942

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75942
Availability details for
Quantity
  • Availability
  • Add to Cart