ENA321763960 Enamine

(4-methylcyclohexyl)(2-methylphenyl)methanamine

  • MDL number MFCD16169976
  • Cas Number : 1250898-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75945

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75945
Availability details for
Quantity
  • Availability
  • Add to Cart