ENA321764338 Enamine

1-methyl-6-oxo-2-phenylpiperidine-3-carboxamide

  • MDL number MFCD18785608
  • Cas Number : 1311317-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75948
Availability details for
Quantity
  • Availability
  • Add to Cart