ENA321764350 Enamine

4-amino-2-(piperidin-1-yl)benzamide

  • MDL number MFCD18785614
  • Cas Number : 1311316-53-6
  • Molecular Weight : 219.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-76019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76019
Availability details for
Quantity
  • Availability
  • Add to Cart