ENA321764352 Enamine

2-(3-methylphenyl)-2-(piperazin-1-yl)acetic acid dihydrochloride

  • MDL number MFCD18838739
  • Cas Number : 1306602-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242...244
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76033

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76033
Availability details for
Quantity
  • Availability
  • Add to Cart