ENA321764391 Enamine

4-(1-amino-2-methylpropan-2-yl)-1$l^{4},4-thiomorpholin-1-one dihydrochloride

  • MDL number MFCD18785630
  • Cas Number : 1311316-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76200

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76200
Availability details for
Quantity
  • Availability
  • Add to Cart