ENA321764392 Enamine

2-(2-aminoethanesulfonyl)-2-methylpropane hydrochloride

  • MDL number MFCD18838762
  • Cas Number : 947664-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76204
Availability details for
Quantity
  • Availability
  • Add to Cart