ENA321764394 Enamine

5-methyl-2-(pyrrolidin-1-yl)benzonitrile

  • MDL number MFCD18785632
  • Cas Number : 1311317-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42...44
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76213

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76213
Availability details for
Quantity
  • Availability
  • Add to Cart