ENA321764397 Enamine

1-(2,6-dimethylpyrimidin-4-yl)-N-propylpiperidin-4-amine dihydrochloride

  • MDL number MFCD18838765
  • Cas Number : 1306605-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76224

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76224
Availability details for
Quantity
  • Availability
  • Add to Cart