ENA321764410 Enamine

[1-(2-chlorophenyl)-2-methylpropyl](methyl)amine hydrochloride

  • MDL number MFCD18838778
  • Cas Number : 1306604-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-76358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76358
Availability details for
Quantity
  • Availability
  • Add to Cart